Merck
[Sesquiterpenoids of Coniogramme maxima].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2012-07-17)
Yunfei Chen, Shoujin Liu, Fei Wang
RÉSUMÉ
To study sesquiterpenoids of Coniogramme maxima. Chemical constituents were separated by chromatography and their structures were identified according to physicochemical property and spectrum data. Fifteen compounds were separated by chromatography technique. Their structures were determined by spectral data, including 10 sesquiterpenoids as (3S)-pteroside D (1), epi-pterosin L (2), pterosin D (3), onitin (4), pterosin Z (5), onitisin (6), onitisin-glucopyranoside (7), onitin-15-O-beta-D-glucopyranoside (8), (2S,3R)-pterosin-L-2'-O-beta-D-glucopyranoside (9) and (3R)-peterosin D-3-O-beta-D-glucopyranoside (10). The other compounds were uracil (11), 3,4-dihydroxybenzaldehyde (12), 5-hydroxymethyl-2-furancarboxaldehyde (13), beta-sitosterol (14) and daucosterol (15). The above 15 compounds are separated from C. maxima for the first time, including 9 compounds being first separated from genus Coniogramme.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Supelco
β-Sitosterol β-D-glucoside, analytical standard