Merck
12-hydroxy-1-azaperylene-limiting case of the ESIPT system: enol-keto tautomerization in S0 and S1 states.
The journal of physical chemistry. A (2012-02-10)
Irena Deperasińska, Daniel T Gryko, Elena Karpiuk, Bolesław Kozankiewicz, Artur Makarewicz, Joanna Piechowska
RÉSUMÉ
Absorption, fluorescence, and fluorescence excitation spectra of 12-hydroxy-1-azaperylene (HAP) and 1-azaperylene were studied in n-alkane matrices at 5 K. Two stable tautomers of HAP, each of them in n-nonane embedded in two sites, were identified and attributed to the enol and keto forms. Theoretical calculations of the energy and vibrational structure of the spectra suggest that tautomer A, with the (0, 0) transition energy at 18,980 ± 10 cm(-1) (and 19,060 ± 10 cm(-1) in the high energy site), should be identified as the keto form, whereas tautomer B, with the (0, 0) energy at 19,200 ± 20 cm(-1) (19,290 ± 20 cm(-1)), as the enol form. Observation of absorption and fluorescence of both tautomeric forms and lack of large Stokes shift of fluorescence of the keto form classify HAP as the limiting case of the excited-state intramolecular proton transfer system.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Nonane, ReagentPlus®, 99%
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Sigma-Aldrich
Nonane, anhydrous, ≥99%
Le tarif et la disponibilité ne sont pas disponibles actuellement.