Merck
Synthesis of pyrazoles via electrophilic cyclization.
The Journal of organic chemistry (2011-07-12)
Metin Zora, Arif Kivrak, Ceyda Yazici
RÉSUMÉ
Electrophilic cyclizations of α,β-alkynic hydrazones by molecular iodine were investigated for the synthesis of 4-iodopyrazoles. α,β-Alkynic hydrazones were readily prepared by the reactions of hydrazines with propargyl aldehydes and ketones. When treated with molecular iodine in the presence of sodium bicarbonate, α,β-alkynic hydrazones underwent electrophilic cyclization to afford 4-iodopyrazoles in good to high yields. Iodocyclization was general for a wide range of α,β-alkynic hydrazones and tolerated the presence of aliphatic, aromatic, heteroaromatic, and ferrocenyl moieties with electron-withdrawing and electron-donating substituents.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
4-Iodopyrazole, 99%