Merck
  • Coenzyme Q10 production by Sphingomonas sp. ZUTE03 with novel precursors isolated from tobacco waste in a two-phase conversion system.
Coenzyme Q10 production by Sphingomonas sp. ZUTE03 with novel precursors isolated from tobacco waste in a two-phase conversion system.
Journal of microbiology and biotechnology (2011-05-28)
Lequan Qiu, Weijian Wang, Weihong Zhong, Li Zhong, Jianjun Fang, Xuanzhen Li, Shijin Wu, Jianmeng Chen
RÉSUMÉ
Coenzyme Q10 (CoQ10) is a widely used supplement in heart diseases treatment or antioxidative dietary. The microbial production of CoQ10 was enhanced by addition of solanesol and novel precursors recovered from waste tobacco. The novel precursors were separated by silica gel and identified as alpha-linolenic acid (LNA) and butylated hydroxytoluene (BHT) based on the effect on CoQ10 production and GC-MS. The effects of novel precursors on CoQ10 production by Sphingomonas sp. ZUTE03 were further evaluated in a two-phase conversion system. The precursor's combination of solanesol (70 mg/l) with BHT (30 mg/l) showed the best effect on the improvement of CoQ10 yield. A maximal CoQ10 productivity (9.5 mg l-1 h-1) was achieved after 8 h conversion, with a molar conversion rate of 92.6% and 92.4% on BHT and solanesol, respectively. The novel precursors, BHT and LNA in crude extracts from waste tobacco leaves, might become potential candidates for application in the industrial production of CoQ10 by microbes.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Solanesol from tobacco, ≥90% (HPLC)