Merck
  • Critical review or scientific opinion paper: arsenosugars--a class of benign arsenic species or justification for developing partly speciated arsenic fractionation in foodstuffs?
Critical review or scientific opinion paper: arsenosugars--a class of benign arsenic species or justification for developing partly speciated arsenic fractionation in foodstuffs?
Analytical and bioanalytical chemistry (2010-10-26)
Jörg Feldmann, Eva M Krupp
RÉSUMÉ
In this opinion paper the toxicokinetic behaviour of arsenosugars is reviewed and compared with that of inorganic arsenic and arsenobetaine. It is concluded that the arsenosugars are similar to inorganic arsenic in terms of metabolite formation and tissue accumulation. As a pragmatic means of generating uniform data sets which adequately represent the toxicity of arsenic in food we recommend reporting partly speciated arsenic concentrations in food commodities in three fractions: i) toxic inorganic arsenic as arsenate (after oxidation); ii) arsenobetaine as established non-toxic arsenic; and iii) potentially toxic arsenic, which includes arsenosugars and other organoarsenicals.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Arsenobetaine, purum p.a., ≥95.0% (NMR)