Merck
Blockade of astrocytic glutamate uptake in rats induces signs of anhedonia and impaired spatial memory.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology (2010-06-10)
Anita J Bechtholt-Gompf, Hali V Walther, Martha A Adams, William A Carlezon, Dost Ongür, Bruce M Cohen
RÉSUMÉ
Mood disorders are associated with regional brain abnormalities, including reductions in glial cell and neuron number, glutamatergic irregularities, and differential patterns of brain activation. Because astrocytes are modulators of neuronal activity and are important in trafficking the excitatory neurotransmitter glutamate, it is possible that these pathologies are interrelated and contribute to some of the behavioral signs that characterize depression and related disorders. We tested this hypothesis by determining whether depressive-like signs were induced by blocking central astrocytic glutamate uptake with the astrocytic glutamate transporter (GLT-1) inhibitor, dihydrokainic acid (DHK), in behavioral tests that quantify aspects of mood, including reward and euthymia/dysthymia: intracranial self-stimulation (ICSS) and place conditioning. We found that DHK elevated ICSS thresholds, a depressive-like effect that could reflect reduced sensitivity to reward (anhedonia) or increased aversion (dysphoria). However, DHK treatment did not establish conditioned place aversions, suggesting that this treatment does not induce dysphoria. To identify the brain regions mediating the behavioral effects of DHK, we examined c-Fos expression in areas implicated in motivation and emotion. DHK increased c-Fos expression in many of these regions. The dentate gyrus of the hippocampus was robustly activated, which led us to explore whether DHK alters hippocampal learning. DHK impaired spatial memory in the MWM. These findings identify disruption of astrocyte glutamate uptake as one component of the complex circuits that mediate anhedonia and cognitive impairment, both of which are common symptoms of depression. These finding may have implications for the etiology of depression and other disorders that share the features of anhedonia and cognitive impairment.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Dihydrokainic acid, ≥98% (HPLC), powder