Merck
Targeting ketone drugs towards transport by the intestinal peptide transporter, PepT1.
Organic & biomolecular chemistry (2009-03-06)
David Foley, Patrick Bailey, Myrtani Pieri, David Meredith
RÉSUMÉ
Thiodipeptide prodrugs of the ketone nabumetone are shown to have affinity for, and be transported by, PepT1 in vitro.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Supelco
Nabumetone, analytical standard
Nabumetone, European Pharmacopoeia (EP) Reference Standard