Merck
Stereospecific high-performance liquid chromatographic determination of tocainide.
Journal of chromatography (1991-05-03)
R A Carr, R T Foster, D Freitag, F M Pasutto
RÉSUMÉ
A sensitive high-performance liquid chromatographic assay was developed for the determination of tocainide enantiomers in plasma. Following extraction of tocainide from plasma, the enantiomers were derivatized with S-(+)-1-(1-naphthyl)ethylisocyanate. The resulting diastereomers were separated and quantified using normal-phase chromatography with fluorescence detection set at 220/345 nm (excitation/emission). The peaks, resolved with a resolution factor greater than 1.5, were free from interference. Linearity was established over the concentration range 0.25-10.0 mg/l for each enantiomer in plasma (r2 greater than 0.998). The inter-assay variability was less than 10% at all concentrations examined. The method can be used to determine the pharmacokinetics of tocainide enantiomers in man.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
(R)-(−)-1-(1-Naphthyl)ethyl isocyanate, 98%