Merck
Chromatographic peak resolution using Microsoft Excel Solver. The merit of time shifting input arrays.
Journal of chromatography. A (2008-09-16)
Purnendu K Dasgupta
RÉSUMÉ
Resolution of overlapped chromatographic peaks is generally accomplished by modeling the peaks as Gaussian or modified Gaussian functions. It is possible, even preferable, to use actual single analyte input responses for this purpose and a nonlinear least squares minimization routine such as that provided by Microsoft Excel Solver can then provide the resolution. In practice, the quality of the results obtained varies greatly due to small shifts in retention time. I show here that such deconvolution can be considerably improved if one or more of the response arrays are iteratively shifted in time.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
3-Nitrophenol, ReagentPlus®, 99%