Merck
Gold-catalyzed cycloisomerization of N-Propargylindole-2-carboxamides: application toward the synthesis of lavendamycin analogues.
Organic letters (2008-07-17)
Dylan B England, Albert Padwa
RÉSUMÉ
A series of N-propargylindole-2-carboxamides were found to undergo a AuCl 3-catalyzed cycloisomerization to give beta-carbolinones in high yield. The corresponding beta-chlorocarboline derivative was prepared and used for Pd(0)-catalyzed cross-coupling chemistry directed toward the synthesis of lavendamycin analogues.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Streptonigrin from Streptomyces flocculus, ≥98%