Merck
  • A microwave spectroscopic and quantum chemical study of propa-1,2-dienyl selenocyanate (H(2)==C==CHSeC[triple bond]N) and cyclopropyl selenocyanate (C(3)H(5)SeC[triple bond]N).
A microwave spectroscopic and quantum chemical study of propa-1,2-dienyl selenocyanate (H(2)==C==CHSeC[triple bond]N) and cyclopropyl selenocyanate (C(3)H(5)SeC[triple bond]N).
Physical chemistry chemical physics : PCCP (2008-07-10)
Rajmund Mokso, Harald Møllendal, Jean-Claude Guillemin
RÉSUMÉ
The microwave spectra of propa-1,2-dienyl selenocyanate, H(2)C==C==CHSeC[triple bond]N, and cyclopropyl selenocyanate, C(3)H(5)SeC[triple bond]N, are reported. The spectra of the ground and two vibrationally excited states of the (80)Se isotopologue and the spectrum of the ground state of the (78)Se isotopologue were assigned for one rotameric form of H(2)C==C[double bond, length as m-dash]CHSeC[triple bond]N. This conformer is characterized by a C-C-Se-C dihedral angle of 129(5) degrees from synperiplanar (0 degrees ) and is shown to be the global minimum of H(2)C[double bond, length as m-dash]C[double bond, length as m-dash]CHSeC[triple bond]N. The spectra of the ground and of three vibrationally excited states of the (80)Se isotopologue, as well as of the ground state of the (78)Se isotopologue of one rotamer of C(3)H(5)SeC[triple bond]N were assigned. This conformer has a H-C-Se-C dihedral angle of 80(4) degrees from synperiplanar and is at least 3 kJ mol(-1) more stable than any other form of the molecule. The microwave study has been augmented by quantum chemical calculations at the B3LYP/6-311+ +G(3df,3pd) and MP2/6-311+ +G(3df,3pd) levels of theory.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Potassium selenocyanate, reagent grade, 97%
Sigma-Aldrich
Potassium selenocyanate, ReagentPlus®, ≥99%