Merck
TAA1-mediated auxin biosynthesis is essential for hormone crosstalk and plant development.
Cell (2008-04-09)
Anna N Stepanova, Joyce Robertson-Hoyt, Jeonga Yun, Larissa M Benavente, De-Yu Xie, Karel Dolezal, Alexandra Schlereth, Gerd Jürgens, Jose M Alonso
RÉSUMÉ
Plants have evolved a tremendous ability to respond to environmental changes by adapting their growth and development. The interaction between hormonal and developmental signals is a critical mechanism in the generation of this enormous plasticity. A good example is the response to the hormone ethylene that depends on tissue type, developmental stage, and environmental conditions. By characterizing the Arabidopsis wei8 mutant, we have found that a small family of genes mediates tissue-specific responses to ethylene. Biochemical studies revealed that WEI8 encodes a long-anticipated tryptophan aminotransferase, TAA1, in the essential, yet genetically uncharacterized, indole-3-pyruvic acid (IPA) branch of the auxin biosynthetic pathway. Analysis of TAA1 and its paralogues revealed a link between local auxin production, tissue-specific ethylene effects, and organ development. Thus, the IPA route of auxin production is key to generating robust auxin gradients in response to environmental and developmental cues.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Indole-3-pyruvic acid, ≥97%