Merck
  • Screening for coumatetralyl in soft drinks by solid-matrix extraction and high-performance liquid chromatography with diode-array detection.
Screening for coumatetralyl in soft drinks by solid-matrix extraction and high-performance liquid chromatography with diode-array detection.
Journal of chromatography (1991-08-16)
A Dimuccio, I Camoni, L Vergori, R Dommarco, D Attard Barbini, F Vergori, A Ausili, A Santilio
RÉSUMÉ
A method was developed for the rapid determination coumatetralyl in cola- and orange-type soft drinks, which includes extraction using solid-matrix column, clean-up by silica cartridge chromatography and reversed-phase high-performance liquid chromatography with diode-array detection. The recovery of coumatetralyl from 50 ml of soft drinks was better than 80% at spiking levels down to 50 micrograms/kg (ppb).
MATÉRIAUX
Référence du produit
Marque
Description du produit
Supelco
Coumatetralyl, PESTANAL®, analytical standard