Merck
Pentobarbital enhances gamma-aminobutyric acid-mediated excitation without altering synaptic plasticity in rat hippocampus.
Anesthesia and analgesia (2007-03-23)
David P Archer, Khanh Q Nguyen, Naaznin Samanani, Sheldon H Roth
RÉSUMÉ
Synaptic plasticity is thought to provide a molecular mechanism for learning and memory. N-methyl-d-aspartate receptor-mediated plasticity requires that N-methyl-d-aspartate receptor activation coincides with postsynaptic depolarizing potentials (DPSP(A)'s). Pentobarbital, in high concentrations, enhances DPSP(A)'s, but high concentrations suppress synaptic plasticity, probably by impairing glutamatergic transmission. Here we tested the hypothesis that low concentrations of pentobarbital can enhance DPSP(A)'s and modify the induction of synaptic plasticity. Studies were performed in vitro on rat hippocampal slices. With glutamate transmission blocked, intracellular recording from CA1 neurons was used to investigate the influence of 5 microM pentobarbital on DPSP(A)'s and neuron excitability evoked by high frequency (100 Hz) stimulation. With glutamate transmission intact, extracellular recording was used to examine the effect of 5 microM pentobarbital on the induction of long-term depression and long-term potentiation of synaptic transmission by conditioning stimuli applied to the Schaffer collateral pathway. High frequency stimulation generated typical DPSP(A)'s that were mediated by gamma-aminobutyric acid(A) receptors and dependent upon HCO3-. Pentobarbital (5 microM) increased the amplitude, but not the width, at half-maximal amplitude of DPSPA's (P < 0.01). Pentobarbital increased the probability of action potential generation during the DPSP(A)'s. Pentobarbital did not alter the induction of long-term depression or long-term potentiation. Despite increasing the amplitude of DPSP(A)'s, 5 microM pentobarbital did not alter the induction of synaptic plasticity by a range of conventional conditioning stimuli. These results do not support the hypothesis that excitatory effects of pentobarbital may alter synaptic plasticity.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Saclofen, solid