Merck
Cholesterol-3-beta, 5-alpha, 6-beta-triol induced genotoxicity through reactive oxygen species formation.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2005-02-22)
Y W Cheng, J J Kang, Y L Shih, Y L Lo, C F Wang
RÉSUMÉ
The mutagenicity of oxysterols, cholesterol-3beta,5alpha,6beta-triol (alpha-Triol), 7-keto-cholesterol (7-Keto) and cholesterol-5alpha,6alpha-epoxide (alpha-Epox) were examined by the Ames method and chromosome aberration test in this study. Only alpha-Triol concentration-dependently caused an increase of bacterial revertants in the absence of metabolic activating enzymes (S9), but not 7-keto and alpha-Epox. The mutagenic effect of alpha-Triol was reduced by the addition of S9. On the other hand, although alpha-Triol significantly induced chromosome aberration in CHO-K1 cells with and without S9. However, the addition of S9 reduced the degree of abnormal structure chromosome compared to without S9 mix. Catalase and superoxide dismutase (SOD) inhibited alpha-Triol induced increase of revertants in Salmonella typhimurium and chromosome aberration frequency in CHO cells, suggesting that reactive oxygen species (ROS) might be involved in the genotoxic effect of alpha-Triol. Treatment with alpha-Triol increased the ROS production in CHO cells, which could be attenuated by catalase and SOD. Results in this study suggested, for the first time that alpha-Triol, causes genotoxic effect in an ROS-dependent manner.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Cholesterol 5α,6α-epoxide
Sigma-Aldrich
Cholesterol 5β,6β-epoxide, ≥98%