Merck
On the mechanisms of oligopeptide reactions in solution and clay dispersion.
Journal of peptide science : an official publication of the European Peptide Society (2005-01-07)
Juraj Bujdák, Bernd Michael Rode
RÉSUMÉ
Mechanisms of the reactions of representative dipeptides (Gly2, Gly-Ala), oligopeptides (Gly3, Gly4) and the polypeptide (poly-Gly)n) in solution and clay suspensions at 85 degrees C were investigated. The reaction products and their yields were analysed and determined by means of HPLC. Interestingly, hydrolysis, where water molecules act as the reactant, was not the main reaction, even for oligopeptides. Formation of cyclic dipeptides prevailed in the reactions of dimers as well as oligopeptides. The breakdown of oligopeptide molecules proceeded via an intramolecular cyclization reaction. For example, the reaction of Gly3 led to the formation of equal amounts of cyclic dipeptide, c(Gly)2 and Gly. The presence of clay (montmorillonite) significantly increased yields in the reactions of dipeptides but it did not have much effect on the reactions of oligopeptides. However, an opposite effect of clay, protection of poly(Gly)n against decomposition, was proven.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Gly-Ala, ≥99.0% (NT)