Merck
  • Toxicosis in dairy cattle exposed to poison hemlock (Conium maculatum) in hay: isolation of Conium alkaloids in plants, hay, and urine.
Toxicosis in dairy cattle exposed to poison hemlock (Conium maculatum) in hay: isolation of Conium alkaloids in plants, hay, and urine.
Journal of veterinary diagnostic investigation : official publication of the American Association of Veterinary Laboratory Diagnosticians, Inc (1992-01-01)
F D Galey, D M Holstege, E G Fisher
RÉSUMÉ
Cattle in two herds developed signs of bloating, increased salivation and lacrimation, depression, respiratory distress, ataxia, and death after ingestion of hay that contained large amounts of poison hemlock (Conium maculatum). Twenty of 30 Angus cows and calves were affected in the first herd (2 died). In the second herd, 5 of 30 Holstein heifers were affected (1 died). The Conium alkaloids, coniine and gamma-coniceine, were quantified in the hay, the plants from the responsible hayfield, and the urine of affected animals.