Merck
Production of antibodies against degradative neoepitopes in aggrecan.
Methods in molecular medicine (2004-07-29)
John S Mort, Peter J Roughley
RÉSUMÉ
The use of synthetic peptides to generate rabbit polyclonal anticatabolic neoepitope antibodies that can be used to study the presence of defined proteolytic cleavage sites in aggrecan is described. Principles of peptide design and methods for preparation and characterization of ovalbumin conjugates are presented along with approaches for the characterization and affinity purification of the resulting antisera. Limitations associated with the use of antipeptide antibodies to study authentic protein neoepitopes are discussed.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Bromoacetic acid N-hydroxysuccinimide ester, ≥95%, powder