Merck
  • Remarkable activity of a novel cyclic seleninate ester as a glutathione peroxidase mimetic and its facile in situ generation from allyl 3-hydroxypropyl selenide.
Remarkable activity of a novel cyclic seleninate ester as a glutathione peroxidase mimetic and its facile in situ generation from allyl 3-hydroxypropyl selenide.
Journal of the American Chemical Society (2002-10-10)
Thomas G Back, Ziad Moussa
RÉSUMÉ
1,2-Oxaselenolane Se-oxide is a novel cyclic seleninate ester that functions as a remarkably efficient glutathione peroxidase mimetic by catalyzing the reduction of tert-butyl hydroperoxide to tert-butyl alcohol in the presence of benzyl thiol. The seleninate ester can be conveniently generated in situ by oxidation of allyl 3-hydroxypropyl selenide with tert-butyl hydroperoxide. Its catalytic activity surpasses that of several other known GPx mimetics containing cyclic selenenamide structures, which were also tested for comparison.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Selenous acid, 98%
Sigma-Aldrich
Selenous acid, 99.999% trace metals basis