Merck
Plasmid stability of recombinant Pseudomonas sp. B13 FR1 pFRC20P in continuous culture.
Biotechnology and bioengineering (1999-04-01)
C Hempel, R W Erb, W Deckwer, V Hecht
RÉSUMÉ
Plasmid stability of recombinant Pseudomonas sp. B13 FR1 pFRC20P, a strain capable of mineralizing 3- and 4-chlorobenzoate and 4-methylbenzoate, was investigated in continuous culture. The hybrid cosmid pFRC20P enables the strain to mineralize 4-methylbenzoate. Rapid plasmid loss was observed under nonselective conditions using 3-chlorobenzoate as the substrate. Plasmid stability decreased with increasing dilution rate. Despite the growth advantage of the generated plasmid free cells a total depletion of plasmid bearing cells was not observed. After approximately 50 generations the fraction of plasmid bearing cells reached a constant level of 10%, which was stably maintained during the next 25 generations. Cells from this stage were used to inoculate a new culture that resulted in a stable level of 50% plasmid bearing cells. By a temporary substrate change to selective conditions (4-methylbenzoate), this level could be further increased to 70%. Literature models on plasmid stability could not be applied to describe the experimental data. Therefore, a new but unstructured model was developed to describe the experimental results. The model is based on the existence of three subpopulations: a plasmid free one, an original plasmid bearing one with a growth disadvantage compared to plasmid free cells, and a second plasmid bearing subpopulation with increased stability that is generated from the original one and has a growth rate comparable to the plasmid free cells.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
p-Toluic acid, 98%