Merck
Anti-austerity agents from Rhizoma et Radix Notopterygii (Qianghuo).
Planta medica (2012-04-21)
Feng Li, Yukari Okamura, Dya Fita Dibwe, Suresh Awale, Shigetoshi Kadota, Yasuhiro Tezuka
RÉSUMÉ
During a search for potent anticancer agents from natural products based on an anti-austerity strategy, we found that a CHCl3 extract of Rhizoma et Radix Notopterygii (Qianghuo), a Chinese crude drug, exhibited strong cytotoxicity against PANC-1 human pancreatic cancer cells, with a PC₅₀ value of 17.5 µg/mL. Further fractionation and purification of this bioactive extract led to the isolation of 19 known compounds. The in vitro preferential cytotoxicity of the isolates was evaluated against two human pancreatic cancer cell lines, PANC-1 and PSN-1. Among the compounds isolated, ostruthin displayed the most potent activity against both PANC-1 (PC₅₀, 7.2 µM) and PSN-1 (PC₅₀, 7.8 µM) cells in nutrient-deprived medium (NDM) and may have induced necrotic nutrient-deprived PANC-1 cell death.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
6-Geranyl-7-hydroxycoumarin, ≥95% (LC/MS-ELSD)