Merck
Drosophila GoLoco-protein Pins is a target of Galpha(o)-mediated G protein-coupled receptor signaling.
Molecular biology of the cell (2009-07-03)
Damir Kopein, Vladimir L Katanaev
RÉSUMÉ
G protein-coupled receptors (GPCRs) transduce their signals through trimeric G proteins, inducing guanine nucleotide exchange on their Galpha-subunits; the resulting Galpha-GTP transmits the signal further inside the cell. GoLoco domains present in many proteins play important roles in multiple trimeric G protein-dependent activities, physically binding Galpha-subunits of the Galpha(i/o) class. In most cases GoLoco binds exclusively to the GDP-loaded form of the Galpha-subunits. Here we demonstrate that the poly-GoLoco-containing protein Pins of Drosophila can bind to both GDP- and GTP-forms of Drosophila Galpha(o). We identify Pins GoLoco domain 1 as necessary and sufficient for this unusual interaction with Galpha(o)-GTP. We further pinpoint a lysine residue located centrally in this domain as necessary for the interaction. Our studies thus identify Drosophila Pins as a target of Galpha(o)-mediated GPCR receptor signaling, e.g., in the context of the nervous system development, where Galpha(o) acts downstream from Frizzled and redundantly with Galpha(i) to control the asymmetry of cell divisions.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Mastoparan-7, ≥97% (HPLC), lyophilized powder