Merck
Simultaneous Measurement of Endothelial Cell Proliferation and Cell Cycle Stage Using Flow Cytometry.
Methods in molecular biology (Clifton, N.J.) (2022-04-23)
Eleanor G Lynam, James A E Lane, Elizabeth J T Finding, Caroline P D Wheeler-Jones
RÉSUMÉ
Endothelial cell proliferation rate is an important indicator of vascular health. Being able to detect the rate of endothelial cell proliferation, or cell cycle disturbances after intervention is a valuable tool for analysing any beneficial or detrimental effects of treatments in vitro. Here, we describe a straightforward flow cytometric-based method of proliferation and cell cycle tracking that can be performed on human endothelial cells in culture over several days.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
β-Endothelial Cell Growth Factor human, β-ECGF, recombinant, expressed in E. coli, suitable for cell culture
Le tarif et la disponibilité ne sont pas disponibles actuellement.