Merck
Galectin-3 is a candidate biomarker for amyotrophic lateral sclerosis: discovery by a proteomics approach.
Journal of proteome research (2010-08-12)
Jian-Ying Zhou, Leila Afjehi-Sadat, Seneshaw Asress, Duc M Duong, Merit Cudkowicz, Jonathan D Glass, Junmin Peng
RÉSUMÉ
The discovery of biomarkers for neurodegenerative diseases will have a major impact on the efficiency of therapeutic clinical trials and may be important for understanding basic pathogenic mechanisms. We have approached the discovery of protein biomarkers for amyotrophic lateral sclerosis (ALS) by profiling affected tissues in a relevant animal model and then validating the findings in human tissues. Ventral roots from SOD1(G93A) "ALS" mice were analyzed by label-free quantitative mass spectrometry, and the resulting data were compared with data for matched samples from nontransgenic littermates and transgenic mice carrying wild-type human SOD1 (SOD1(WT)). Of 1299 proteins, statistical inference of the data in the three groups identified 14 proteins that were dramatically altered in the ALS mice compared with the two control groups. The protein galectin-3 emerged as a lead biomarker candidate on the basis of its differential expression as assessed by immunoblot and immunocytochemistry in SOD1(G93A) mice as compared to controls and because it is a secreted protein that could potentially be measured in human biofluids. Spinal cord tissue from ALS patients also exhibited increased levels of galectin-3 when compared to controls. Further measurement of galectin-3 in cerebrospinal fluid samples showed that ALS patients had approximately twice as much galectin-3 as normal and disease controls. These results provide the proof of principle that biomarker identification in relevant and well-controlled animal models can be translated to human disease. The challenge is to validate our biomarker candidate proteins as true biomarkers for ALS that will be useful for diagnosis and/or monitoring disease activity in future clinical trials.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Apolipoprotein E, Human Plasma, Very Low-Density Lipoprotein, Native apolipoprotein E from human plasma. A major component of VLDL and IDL. Serves as a major cholesterol carrier. Essential for the normal catabolism of triglyceride-rich lipoprotein constituents.