Merck
Carcinoembryonic antigen as a specific glycoprotein ligand of rBC2LCN lectin on pancreatic ductal adenocarcinoma cells.
Cancer science (2021-06-12)
Tomoaki Furuta, Tatsuya Oda, Kayo Kiyoi, Ozawa Yusuke, Sota Kimura, Ko Kurimori, Yoshihiro Miyazaki, Yang Yu, Kinji Furuya, Yoshimasa Akashi, Osamu Shimomura, Hiroaki Tateno
RÉSUMÉ
The rBC2LCN lectin, known as a stem cell marker probe that binds to an H type 3 fucosylated trisaccharide motif, was recently revealed to also bind to pancreatic ductal adenocarcinoma (PDAC) cells. A lectin-drug conjugate was generated by fusing rBC2LCN with a cytocidal toxin, and it showed a strong anticancer effect in in vitro and in vivo PDAC models. However, it is unclear which molecules are carrier proteins of rBC2LCN on PDAC cells. In this study, we identified a rBC2LCN-positive glycoprotein expressed in PDAC. Tumor lysates of PDAC patient-derived xenografts (PDXs) were coprecipitated with rBC2LCN lectin and analyzed by liquid chromatography-mass spectrometry. A total of 343 proteins were initially identified. We used a web-based database to select five glycoproteins and independently evaluated their expression in PDAC by immunohistochemistry (IHC). Among them, we focused on carcinoembryonic antigen 5 (CEA) as the most cancer-specific carrier protein in PDAC, as it showed the most prominent difference in expression rate between PDAC cells (74%) and normal pancreatic duct cells (0%, P > .0001). rBC2LCN lectin and CEA colocalization in PDAC samples was confirmed by double-staining analysis. Furthermore, rBC2LCN-precipitated fractions were blotted with an anti-CEA polyclonal antibody (pAb), and CEA pAb-precipitated fractions were blotted with rBC2LCN lectin. The results demonstrate that CEA is in fact a ligand of rBC2LCN lectin.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
CelLytic MEM Protein Extraction Kit