Merck
Analgesic and Antipyretic Activities of Ethyl Acetate Fraction Tablet of Andrographis paniculata in Animal Models.
Evidence-based complementary and alternative medicine : eCAM (2021-03-23)
Hilkatul Ilmi, Irfan Rayi Pamungkas, Lidya Tumewu, Achmad Fuad Hafid, Aty Widyawaruyanti
RÉSUMÉ
To determine the analgesic and antipyretic activities of a tablet derived from Andrographis paniculata ethyl acetate fraction (AS201-01) in animal models. The tablet derived from AS201-01 contains an equivalent of 35 mg andrographolide per tablet. Analgesic activity was determined using an acetic acid-induced writhing test on adult male mice. A writhe was recorded by a stopwatch and was defined as the stretching of the abdomen and/or stretching of at least one hind limb. For the determination of antipyretic activity, pyrexia was induced by subcutaneous injection of 15% w/v Brewer's yeast into adult male rats. Rectal temperature was monitored at 1, 2, 3, and 4 hours after treatment. The results showed that the AS201-01 tablet had analgesic and antipyretic activity. In the acetic acid-induced writhing model, AS201-01 tablet exhibited significant analgesic effect with a 66.73% reduction in writhing response at a dose of 50 mg andrographolide/kg body weight compared to the negative control group. The tablet also showed a significant antipyretic effect. The maximum antipyretic effect was observed after the third hour of administration of the AS201-01 tablet at a dose of 100 mg andrographolide/kg body weight. Tablet of Andrographis paniculata ethyl acetate fraction (AS201-01) exhibited analgesic and antipyretic activities.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Yeast, powder, slightly brown