Merck
Synthesis of a C20-C26 segment of superstolide A by addition of a chiral allenylzinc reagent to (R)-N-boc alaninal.
Organic letters (2005-04-09)
James A Marshall, James J Mulhearn
RÉSUMÉ
[reaction: see text] Additions of chiral allenylzinc and indium reagents to N-Boc alaninal were examined as a possible route to a C20-C26 segment of superstolide A. Allenylzinc reagents, prepared in situ by palladiozincation of (R)- and (S)-5-pivalyloxy-3-butyn-2-ol mesylate, showed excellent reagent control to afford the anti,syn and anti,anti diastereomers as the nearly exclusive adducts.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Boc-L-alaninal, ≥98%