- Role of JNK activation in apoptosis: a double-edged sword.
Role of JNK activation in apoptosis: a double-edged sword.
Cell research (2005-02-03)
Jing Liu, Anning Lin
PMID15686625
RÉSUMÉ
JNK is a key regulator of many cellular events, including programmed cell death (apoptosis). In the absence of NF-kB activation, prolonged JNK activation contributes to TNF-a induced apoptosis. JNK is also essential for UV induced apoptosis. However, recent studies reveal that JNK can suppress apoptosis in IL-3-dependent hematopoietic cells via phosphorylation of the proapoptotic Bcl-2 family protein BAD. Thus, JNK has pro- or antiapoptotic functions, depending on cell type, nature of the death stimulus, duration of its activation and the activity of other signaling pathways.