Merck
Horseradish peroxidase-catalyzed oxidation of acetaminophen to intermediates that form polymers or conjugate with glutathione.
Molecular pharmacology (1986-02-01)
D W Potter, D W Miller, J A Hinson
RÉSUMÉ
Horseradish peroxidase catalyzed the polymerization of acetaminophen. Addition of reduced glutathione (GSH) to reaction mixtures resulted in decreased polymerization and formation of minor amounts of GSH-acetaminophen conjugates. The conjugates were identified as 3-(glutathion-S-yl)acetaminophen and 3-(glutathion-S-yl)diacetaminophen. Horseradish peroxidase also catalyzed polymerization of synthetic 3-(glutathion-S-yl)acetaminophen to a dimer conjugate. In contrast to acetaminophen, 3-(glutathion-S-yl)acetaminophen oxidation was slowly catalyzed by horseradish peroxidase. However, in reaction mixtures containing equimolar concentrations of acetaminophen and synthetic 3-(glutathion-S-yl)acetaminophen, the formation of 3-(glutathion-S-yl)diacetaminophen and 3-(diglutathion-S-yl)diacetaminophen was rapid and accounted for approximately 95% of the products, whereas acetaminophen polymers accounted for only 5% of the products. These findings suggest that horseradish peroxidase catalyzed the one-electron oxidation of acetaminophen to N-acetyl-p-benzosemiquinone imine which preferentially polymerized rather than reacted with GSH. N-Acetyl-p-benzosemiquinone imine may also oxidize 3-(glutathion-S-yl)acetaminophen to form acetaminophen and 3-(glutathion-S-yl)-N-acetyl-p-benzosemiquinone imine. The data indicate that once this conjugate radical is formed it reacts with either N-acetyl-p-benzosemiquinone minine or 3-(glutathion-S-yl)-N-acetyl-p-benzosemiquinone imine via a radical termination mechanism.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Peroxydase from horseradish, Highly stabilized, essentially salt-free, lyophilized powder, 200-300 units/mg solid (using pyrogallol)