Merck
Enzymatic production of 5'-inosinic acid by AMP deaminase from a newly isolated Aspergillus oryzae.
Food chemistry (2016-09-07)
Shubo Li, Leitao Chen, Yangjun Hu, Guohui Fang, Mouming Zhao, Yuan Guo, Zongwen Pang
RÉSUMÉ
5'-adenylic acid deaminase (AMP deaminase), an important enzyme for the food industry, can catalyze the irreversible hydrolysis of adenosine monophosphate (AMP) to inosine monophosphate (IMP) and ammonia. In this study, a new strain was screened that efficiently produces 3191.6U/g of AMP deaminase at 32°C. After purification, the optimal temperature and pH of the AMP deaminase were found to be 40°C and 6.0, respectively, but it was partially inhibited by Fe(3+), Cu(2+), Al(3+), and Zn(2+). With amplification of the AMP deaminase production system, 6mL of crude enzyme could produce 2.00mg/g of IMP from 2.04mg/g of dried yeast with an 84.8% molar yield after 40min. These results provide a new insight into AMP deaminase production and offer a potential platform for producing 5'-IMP.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Inosine 5′-monophosphate disodium salt hydrate, from yeast, Grade III, ≥98% (HPLC)
Le tarif et la disponibilité ne sont pas disponibles actuellement.