Merck
JQ1, an inhibitor of the epigenetic reader BRD4, suppresses the bidirectional MYC-AP4 axis via multiple mechanisms.
Oncology reports (2015-11-18)
Sung Kyung Choi, Seong Hwi Hong, Hyuk Soon Kim, Chan Young Shin, Suk Woo Nam, Wahn Soo Choi, Jeung-Whan Han, Jueng Soo You
RÉSUMÉ
Bromodomain and extra-terminal domain (BET) family proteins are representative epigenetic modulators that read acetylated lysine residues and transfer cellular signals. Recently, the BET protein inhibitor JQ1 was developed and has been extensively studied in many cancer cell types. We demonstrated that JQ1 effectively suppressed the MYC-AP4 axis and induced antitumorigenic effects by targeting a bidirectional positive loop between MYC and AP4 which was first proposed in the present study. MYC and AP4 are the direct targets of BRD4, as demonstrated by chromatin immunoprecipitation (ChIP) assay and BRD4 loss-of-function experiments. Although inhibition of the MYC/MAC dimer suppressed AP4, the efficacy of suppression was not as effective as BRD4 inhibition. Notably, AP4 loss-of-function studies demonstrated that AP4 is a major critical target of JQ1 and that MYC is a novel downstream target of AP4, as demonstrated by AP4 binding to the MYC promoter. Taken together, our results suggest that the epigenetic reader BRD4 is a key mediator of the activated MYC-AP4 axis, which supports the possibility that targeting BET protein is a novel therapeutic strategy for MYC-AP4 axis-activated cancers.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anti-TFAP4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution