Merck
Spatial and temporal tracking of cardiac exosomes in mouse using a nano-luciferase-CD63 fusion protein.
Communications biology (2020-03-12)
Weijia Luo, Yuan Dai, Zhishi Chen, Xiaojing Yue, Kelsey C Andrade-Powell, Jiang Chang
RÉSUMÉ
Exosomes are secreted extracellular vesicles with lipid bilayer membranes. They are emerging as a new category of messengers that facilitate cross-talk between cells, tissues, and organs. Thus, a critical demand arises for the development of a sensitive and non-invasive tracking system for endogenous exosomes. We have generated a genetic mouse model that meets this goal. The Nano-luciferase (NanoLuc) reporter was fused with the exosome surface marker CD63 for exosome labeling. The cardiomyocyte-specific αMHC promoter followed by the loxP-STOP-loxP cassette was engineered for temporal and spatial labeling of exosomes originated from cardiomyocytes. The transgenic mouse was bred with a tamoxifen-inducible Cre mouse (Rosa26Cre-ERT2) to achieve inducible expression of CD63NanoLuc reporter. The specific labeling and tissue distribution of endogenous exosomes released from cardiomyocytes were demonstrated by luciferase assay and non-invasive bioluminescent live imaging. This endogenous exosome tracking mouse provides a useful tool for a range of research applications.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Thrombin, Bovine