Merck
  • Hyperthermia exacerbates the acute effects of psychoactive substances on neuronal activity measured using microelectrode arrays (MEAs) in rat primary cortical cultures in vitro.
Hyperthermia exacerbates the acute effects of psychoactive substances on neuronal activity measured using microelectrode arrays (MEAs) in rat primary cortical cultures in vitro.
Toxicology and applied pharmacology (2020-04-23)
Anne Zwartsen, Laura Hondebrink, Dylan W de Lange, Remco H S Westerink
RÉSUMÉ
Hyperthermia is a well-known, potentially life-threatening, side effect of stimulant psychoactive substances that worsens the neurological outcome of hospitalized patients. However, current in vitro methods to assess the hazard of psychoactive substances do not account for hyperthermia. Therefore, this study determined the potency of five psychoactive substances (cocaine, MDMA (3,4-methylenedioxymethamphetamine), methamphetamine, 3-MMC (3-methylmethcathinone) and TFMPP (3-trifluoromethylphenylpiperazine)) to affect neuronal activity at physiological and hyperthermic conditions. Neuronal activity of rat cortical cultures grown on microelectrode arrays (MEAs) was recorded at 37 °C before exposure. Following 30 min and 4.5 h drug exposure (1-1000 μM) at 37 °C or 41 °C, neuronal activity was measured at either 37 °C or 41 °C. Without drug exposure, hyperthermia induced a modest decrease in neuronal activity. Following acute (30 min) exposure at 37 °C, all drugs concentration-dependently inhibited neuronal activity. Increasing the temperature to 41 °C significantly exacerbated the reduction of neuronal activity ~ 2-fold for all drugs compared to 37 °C. Prolonged (4.5 h) exposure at 41 °C decreased neuronal activity comparable to 37 °C. Neuronal activity (partly) recovered following drug exposure at both temperatures, although recovery from exposure at 41 °C was less pronounced for most drugs. None of the exposure conditions affected viability. Since acute exposure at hyperthermic conditions exacerbates the decrease in neuronal activity induced by psychoactive substances, effects of hyperthermia should be included in future hazard assessment of illicit drugs and new psychoactive substances (NPS).