Merck
  • Two Nucleoporin98 homologous genes jointly participate in the regulation of starch degradation to repress senescence in Arabidopsis.
Two Nucleoporin98 homologous genes jointly participate in the regulation of starch degradation to repress senescence in Arabidopsis.
BMC plant biology (2020-06-27)
Long Xiao, Shanshan Jiang, Penghui Huang, Fulu Chen, Xu Wang, Zhiyuan Cheng, Yuchen Miao, Liangyu Liu, Iain Searle, Chunyan Liu, Xiao-Xia Wu, Yong-Fu Fu, Qingshan Chen, Xiao-Mei Zhang
RÉSUMÉ
Starch is synthesized during daylight for temporary storage in leaves and then degraded during the subsequent night to support plant growth and development. Impairment of starch degradation leads to stunted growth, even senescence and death. The nuclear pore complex is involved in many cellular processes, but its relationship with starch degradation has been unclear until now. We previously identified that two Nucleoporin98 genes (Nup98a and Nup98b) redundantly regulate flowering via the CONSTANS (CO)-independent pathway in Arabidopsis thaliana. The double mutant also shows severe senescence phenotypes. We find that Nucleoporin 98 participates in the regulation of sugar metabolism in leaves and is also involved in senescence regulation in Arabidopsis. We show that Nup98a and Nup98b function redundantly at different stages of starch degradation. The nup98a-1 nup98b-1 double mutant accumulates more starch, showing a severe early senescence phenotype compared to wild type plants. The expression of marker genes related to starch degradation is impaired in the nup98a-1 nup98b-1 double mutant, and marker genes of carbon starvation and senescence express their products earlier and in higher abundance than in wild type plants, suggesting that abnormalities in energy metabolism are the main cause of senescence in the double mutant. Addition of sucrose to the growth medium rescues early senescence phenotypes of the nup98a-1 nup98b-1 mutant. Our results provide evidence for a novel role of the nuclear pore complex in energy metabolism related to growth and development, in which Nup98 functions in starch degradation to control growth regulation in Arabidopsis.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Murashige and Skoog Basal Salt Mixture (MS), powder, suitable for plant cell culture