- Iron-catalyzed intramolecular allylic C-H amination.
Iron-catalyzed intramolecular allylic C-H amination.
Journal of the American Chemical Society (2012-01-21)
Shauna M Paradine, M Christina White
PMID22260649
RÉSUMÉ
A highly selective C-H amination reaction under iron catalysis has been developed. This novel system, which employs an inexpensive, nontoxic [Fe(III)Pc] catalyst (typically used as an industrial ink additive), displays a strong preference for allylic C-H amination over aziridination and all other C-H bond types (i.e., allylic > benzylic > ethereal > 3° > 2° ≫ 1°). Moreover, in polyolefinic substrates, the site selectivity can be controlled by the electronic and steric character of the allylic C-H bond. Although this reaction is shown to proceed via a stepwise mechanism, the stereoretentive nature of C-H amination for 3° aliphatic C-H bonds suggests a very rapid radical rebound step.