Merck
Pd(II)-catalyzed carbonylation of C(sp3)-H bonds: a new entry to 1,4-dicarbonyl compounds.
Journal of the American Chemical Society (2010-11-19)
Eun Jeong Yoo, Masayuki Wasa, Jin-Quan Yu
RÉSUMÉ
Pd(II)-catalyzed β-C(sp(3))-H carbonylation of N-arylamides under CO (1 atm) has been achieved. Following amide-directed C(sp(3))-H cleavage and insertion of CO into the resulting [Pd(II)-C(sp(3))] bond, intramolecular C-N reductive elimination gave the corresponding succinimides, which could be readily converted to 1,4-dicarbonyl compounds. This method was found to be effective with substrates containing α-hydrogen atoms and could be applied to effect methylene C(sp(3))-H carbonylation of cyclopropanes.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Yu-Wasa Auxiliary, 97%