Merck
  • Hippocampal HECT E3 ligase inhibition facilitates consolidation, retrieval, and reconsolidation, and inhibits extinction of contextual fear memory.
Hippocampal HECT E3 ligase inhibition facilitates consolidation, retrieval, and reconsolidation, and inhibits extinction of contextual fear memory.
Neurobiology of learning and memory (2019-12-11)
Jadier Redondo, Bruno Popik, Mirelle Casagrande, Mateus Oliveira Silva, Jorge Alberto Quillfeldt, Lucas de Oliveira Alvares, Tadeu Mello E Souza
RÉSUMÉ
Ubiquitination is involved in synaptic plasticity and memory, but the involvement of HECT E3 ligases in these processes has not yet been established. Here, we bilaterally infused heclin, a specific inhibitor of some of these ligases, into the dorsal hippocampus of male Wistar rats that were trained in a contextual fear conditioning. Heclin improved short-term memory, consolidation, retrieval, and reconsolidation when administered immediately post training, prior to testing, or after memory reactivation, respectively. In addition, it impaired memory extinction when administered prior to a long reactivation session. Heclin infusion was also tested for locomotor activity and anxiety-like behavior in a circular arena, but no effect was seen. Taken together, these results indicate that HECT E3 ligases are involved in the modulation of fear memory.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Heclin, ≥98% (HPLC)