Merck
Thermotropic and barotropic phase behavior of phosphatidylcholine bilayers.
International journal of molecular sciences (2013-01-26)
Hitoshi Matsuki, Masaki Goto, Kaori Tada, Nobutake Tamai
RÉSUMÉ
Bilayers formed by phospholipids are frequently used as model biological membranes in various life science studies. A characteristic feature of phospholipid bilayers is to undergo a structural change called a phase transition in response to environmental changes of their surroundings. In this review, we focus our attention on phase transitions of some major phospholipids contained in biological membranes, phosphatidylcholines (PCs), depending on temperature and pressure. Bilayers of dipalmitoylphosphatidylcholine (DPPC), which is the most representative lipid in model membrane studies, will first be explained. Then, the bilayer phase behavior of various kinds of PCs with different molecular structures is revealed from the temperature-pressure phase diagrams, and the difference in phase stability among these PC bilayers is discussed in connection with the molecular structure of the PC molecules. Furthermore, the solvent effect on the phase behavior is also described briefly.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Avanti
16:0-14:0 PC, 1-palmitoyl-2-myristoyl-sn-glycero-3-phosphocholine, powder