Merck
IL-6 and IL-8 are involved in JMJD2A-regulated malignancy of ovarian cancer cells.
Archives of biochemistry and biophysics (2020-03-17)
Haiyu Zhang, Zichao Wang, Fengxia Wang, Chengdong Wang, Hui Zhang
RÉSUMÉ
Emerging evidence shows that histone modification and its related regulators are involved in the progression and chemoresistance of ovarian cancer (OC) cells. Our present study found that the expression of Jumonji C domain-containing 2A (JMJD2A), while not JMJD2B or JMJD2C, is increased in OC cells and tissues as compared with that in their corresponding controls. Knockdown of JMJD2A can decrease proliferation while increase cisplatin (CDDP) sensitivity of OC cells. By screening the expression of cytokines involved in the progression of ovarian cancer, we found that knockdown of JMJD2A can inhibit the expression of interleukin-6 (IL-6) and IL-8 in ovarian cancer cells. Recombinant IL-6 (rIL-6) and rIL-8 can attenuate si-JMJD2A-suppressed malignancy of OC cells. Mechanistically, JMJD2A can directly bind with the promoter of IL-6 to trigger its transcription. For IL-8, JMJD2A can increase it mRNA stability in OC cells. Collectively, we revealed that JMJD2A can trigger the malignancy of OC cells via upregulation of IL-6 and IL-8. It suggested that JMJD2A might be a potential target for OC treatment and therapy.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
MISSION® esiRNA, targeting human KDM4A