Merck
Distinct regulatory ribosomal ubiquitylation events are reversible and hierarchically organized.
eLife (2020-02-06)
Danielle M Garshott, Elayanambi Sundaramoorthy, Marilyn Leonard, Eric J Bennett
RÉSUMÉ
Activation of the integrated stress response (ISR) or the ribosome-associated quality control (RQC) pathway stimulates regulatory ribosomal ubiquitylation (RRub) on distinct 40S ribosomal proteins, yet the cellular role and fate of ubiquitylated proteins remain unclear. We demonstrate that uS10 and uS5 ubiquitylation are dependent upon eS10 or uS3 ubiquitylation, respectively, suggesting that a hierarchical relationship exists among RRub events establishing a ubiquitin code on ribosomes. We show that stress dependent RRub events diminish after initial stimuli and that demodification by deubiquitylating enzymes contributes to reduced RRub levels during stress recovery. Utilizing an optical RQC reporter we identify OTUD3 and USP21 as deubiquitylating enzymes that antagonize ZNF598-mediated 40S ubiquitylation and can limit RQC activation. Critically, cells lacking USP21 or OTUD3 have altered RQC activity and delayed eS10 deubiquitylation indicating a functional role for deubiquitylating enzymes within the RQC pathway.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Anti-ZNF598 antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-USP21 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution