Merck
Latex proteins from Calotropis procera: Toxicity and immunological tolerance revisited.
Chemico-biological interactions (2017-07-16)
Camila F Bezerra, Érika F Mota, Ana Claudia M Silva, Adriana R Tomé, Maria Z R Silva, Daniel de Brito, Camila T M N Porfírio, Ariclécio C Oliveira, José V Lima-Filho, Márcio V Ramos
RÉSUMÉ
Many thousands of plants are disseminated worldwide in traditional and folk medicines based on the belief that their leaves, roots, seeds, bark or secretions, when adequately handled, can treat, alleviate or ameliorate numerous disease symptoms. Calotropis procera (Apocynaceae) is a popular medicinal plant and the claims of this shrub's phytomedicinal properties have been scientifically validated. In this study, further prospects towards the in vivo toxicity and oral immunological tolerance of phytomodulatory proteins isolated from the latex of C. procera are reported. Acute toxicity was determined in mice by oral and intraperitoneal administration of latex proteins (LP) and was followed behavioral, hematological and histological analyses. Oral immunological tolerance to LP was assessed by intraperitoneal immunization in mice that had received LP orally before. Animals given 5000 mg/kg orally exhibited only discrete behavioral alterations and augmentation of monocytes. Death was not notified 14 days after exposure. However, all animals receiving LP 150 mg/kg by i.p. died in 1 h. Death (20%) was documented when LP (75 mg/kg) was given in the peritoneum and signs of harmful effects were observed in the survivors (80%). Oral immunological tolerance was observed in animals previously given LP orally, when they were further immunized/challenged with peritoneal exposure to different doses of LP. This was confirmed by the lowering of IgE and IgG in the serum, IL-4 and IFN-γ in spleen homogenates and the absence of anaphylaxis signs. It is therefore concluded that LP exhibited quite discrete adverse effects when orally administrated at higher concentrations and this route of administration did not stimulate adverse immunological reactions. Instead it was observed immunological tolerance. The present study contributes very important information concerning the safe use of C. procera as a phytotherapeutic agent.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Mouse IgE ELISA Kit
Le tarif et la disponibilité ne sont pas disponibles actuellement.