Merck
Cytoplasmic dsRNA induces the expression of OCT3/4 and NANOG mRNAs in differentiated human cells.
The Journal of biological chemistry (2019-10-17)
Guanming Wang, Takahisa Kouwaki, Kazuki Mugikura, Masaaki Okamoto, Hiromi Takaki, Kenji Funami, Tsukasa Seya, Hiroyuki Oshiumi
RÉSUMÉ
Cytoplasmic dsRNA is recognized by RNA helicase RIG-I (RIG-I) and melanoma differentiation-associated protein 5 (MDA5), triggering induction of the innate immune response via the mitochondrial antiviral signaling protein (MAVS). In contrast, extracellular dsRNA is internalized into endosomes and recognized by Toll-like receptor 3 (TLR3), which triggers signaling via the Toll-like receptor adaptor molecule 1 (TICAM-1). Poly(I:C) is a synthetic dsRNA analog and increases the expression of octamer-binding protein 3/4 (OCT3/4), NANOG, and SRY-box (SOX) mRNAs during pluripotency induction. However, the mechanism underlying this increase is unclear. Here, we focused on the mechanism of poly(I:C)-induced expression of stem cell-specific genes in human somatic cells. Addition of poly(I:C) to human fibroblast culture medium did not increase OCT3/4 mRNA expression, but poly(I:C) transfection markedly increased OCT3/4 expression and induced nuclear localization of the OCT3/4 protein, implying that not TLR3, but RIG-I and MDA5 are required for OCT3/4 expression. Moreover, although cytoplasmic dsRNA increased OCT3/4 mRNA, cytoplasmic dsDNAs, such as salmon sperm DNA and poly(dA:dT), did not. Interestingly, the expression of NANOG, SOX2, Krüppel-like factor 4 (KLF4), and proto-oncogene c-Myc was also increased by cytoplasmic dsRNA. Of note, siRNAs that silenced MAVS and interferon regulatory factor 1 (IRF1) expression reduced OCT3/4 levels after stimulation with poly(I:C); however, an NF-κB inhibitor and siRNA-mediated knockdown of proto-oncogene c-Jun did not significantly reduce the mRNA levels. We conclude that cytoplasmic dsRNA increases the expression of stem cell-specific genes in human somatic cells in a MAVS- and IRF1-dependent manner.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
MISSION® esiRNA, targeting human STAT2