Merck
A Novel Systems-Biology Algorithm for the Analysis of Coordinated Protein Responses Using Quantitative Proteomics.
Molecular & cellular proteomics : MCP (2016-02-20)
Fernando García-Marqués, Marco Trevisan-Herraz, Sara Martínez-Martínez, Emilio Camafeita, Inmaculada Jorge, Juan Antonio Lopez, Nerea Méndez-Barbero, Simón Méndez-Ferrer, Miguel Angel Del Pozo, Borja Ibáñez, Vicente Andrés, Francisco Sánchez-Madrid, Juan Miguel Redondo, Elena Bonzon-Kulichenko, Jesús Vázquez
RÉSUMÉ
The coordinated behavior of proteins is central to systems biology. However, the underlying mechanisms are poorly known and methods to analyze coordination by conventional quantitative proteomics are still lacking. We present the Systems Biology Triangle (SBT), a new algorithm that allows the study of protein coordination by pairwise quantitative proteomics. The Systems Biology Triangle detected statistically significant coordination in diverse biological models of very different nature and subjected to different kinds of perturbations. The Systems Biology Triangle also revealed with unprecedented molecular detail an array of coordinated, early protein responses in vascular smooth muscle cells treated at different times with angiotensin-II. These responses included activation of protein synthesis, folding, turnover, and muscle contraction - consistent with a differentiated phenotype-as well as the induction of migration and the repression of cell proliferation and secretion. Remarkably, the majority of the altered functional categories were protein complexes, interaction networks, or metabolic pathways. These changes could not be detected by other algorithms widely used by the proteomics community, and the vast majority of proteins involved have not been described before to be regulated by AngII. The unique capabilities of The Systems Biology Triangle to detect functional protein alterations produced by the coordinated action of proteins in pairwise quantitative proteomics experiments make this algorithm an attractive choice for the biological interpretation of results on a routine basis.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anticorps anti-ténascine, Chemicon®, from rabbit
Le tarif et la disponibilité ne sont pas disponibles actuellement.