Merck
Long-term retinal cone rescue using a capsid mutant AAV8 vector in a mouse model of CNGA3-achromatopsia.
PloS one (2017-11-14)
Xufeng Dai, Ying He, Hua Zhang, Yangyang Zhang, Yan Liu, Muran Wang, Hao Chen, Ji-Jing Pang
RÉSUMÉ
Adeno-associated virus (AAV) vectors are important gene delivery tools for the treatment of many recessively inherited retinal diseases. For example, a wild-type (WT) AAV5 vector can deliver a full-length Cnga3 (cyclic nucleotide-gated channel alpha-3) cDNA to target cells of the cone photoreceptor function loss 5 (cpfl5) mouse, a spontaneous animal model of achromatopsia with a Cnga3 mutation. Gene therapy restores cone-mediated function and blocks cone degeneration in the mice. However, since transgene expression delivered by an AAV vector shows relatively short-term effectiveness, this cannot be regarded as a very successful therapy. AAV2 and AAV8 vectors with capsid mutations have significantly enhanced transduction efficiency in retinas compared to WT AAV controls. In this study, AAV8 (Y447, 733F+T494V)-treated cpfl5 retinas showed greater preservation of short-term cone electroretinogram (ERG) responses than AAV8 (Y447, 733F)- or AAV2 (Y272, 444, 500, 730F+T491V)-mediated treatments. To explore the long-term rescue effect, AAV8 (Y447, 733F+T494V)-treated cpfl5 retinas were evaluated at 9 months following postnatal day 14 (P14) treatment. Rescued ERG responses in the cones of treated cpfl5 eyes decreased with increasing age, but still maintained more than 60% of the WT mouse responses at the oldest time point examined. Expression of CNGA3 and M/S-opsins was maintained in cone outer segments of the treated cpfl5 eyes and was equal to expression in age-matched WT retinas. Near-normal cone-mediated water maze behavior was observed in the treated cpfl5 mice. As these are the longest follow-up data reported thus far, AAV8 with capsid Y-F and T-V mutations may be one of the most effective AAV vectors for long-term treatment in a naturally occurring mouse model of CNGA3 achromatopsia.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, Cy3 conjugate, Species Adsorbed, 1.5 mg/mL, Chemicon®