Merck
PILAR is a novel modulator of human T-cell expansion.
Blood (2008-06-14)
Eduardo Huarte, Juan R Cubillos-Ruiz, Yolanda C Nesbeth, Uciane K Scarlett, Diana G Martinez, Xavier A Engle, William F Rigby, Patricia A Pioli, Paul M Guyre, Jose R Conejo-Garcia
RÉSUMÉ
Robust T-cell responses without autoimmunity are only possible through a fine balance between activating and inhibitory signals. We have identified a novel modulator of T-cell expansion named proliferation-induced lymphocyte-associated receptor (PILAR). Surface PILAR is markedly up-regulated on CD4 and, to a lesser extent, on CD8 T cells on T-cell receptor engagement. In absence of CD28 costimulation, PILAR signaling through CD161 supports CD3 antibody-dependent and antigen-specificT-cell proliferation by increasing the expression of antiapoptotic Bcl-xL and induces secretion of T helper type 1 cytokines. These effects are abrogated by PILAR blockade with specific antibodies, which decrease surface levels of CD28. In contrast, PILAR induces apoptotic death on naive and early activated T cells if CD161 engagement is blocked. PILAR is expressed by approximately 7% to 10% of CD4 T cells in 2 samples of inflammatory synovial fluid, suggesting a potential role in the pathogenesis of joint inflammation. In addition, in the ovarian cancer microenvironment, effector T cells express PILAR, but not CD161, although expression of both can be augmented ex vivo. Our results indicate that PILAR plays a central role in modulating the extent of T-cell expansion. Manipulation of PILAR signaling may be important for treatment of autoimmune diseases and cancer.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anti-CD28 Antibody, clone 15E8, clone 15E8, Chemicon®, from mouse