Merck
Two-electron oxygen reduction on fullerene C60-carbon nanotubes covalent hybrid as a metal-free electrocatalyst.
Scientific reports (2019-09-26)
Aliyeh Hasanzadeh, Alireza Khataee, Mahmoud Zarei, Yifeng Zhang
RÉSUMÉ
Nanocarbon materials are considered to be active for electrochemical oxygen reduction reaction (ORR) for hydrogen peroxide (H2O2) synthesis. In the present work, a new type of fullerene 60 (C60)-carbon nanotubes (CNTs) hybrid with covalently attached C60 onto outer surface of CNTs was synthesized. The structure of C60-CNT hybrid was confirmed by physical and chemical characterizations and its conformation is proposed featuring the covalent incorporation of CNTs and C60 derivative. C60-CNT hybrid showed high efficiencies on electro-generating H2O2, owing to huge surface area and intermolecular electron-transfer in the hybrid structure. A high H2O2 production rate of 4834.57 mg L-1 h-1 (426.58 mmol L-1) was achieved at - 0.2 V vs saturated calomel electrode (SCE).
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Acide 4-chlorobenzoïque, 99%