Merck
  • Lysolecithins as endothelium-dependent vascular smooth muscle relaxants that differ from endothelium-derived relaxing factor (nitric oxide)
Lysolecithins as endothelium-dependent vascular smooth muscle relaxants that differ from endothelium-derived relaxing factor (nitric oxide)
Proceedings of the National Academy of Sciences of the United States of America (1988-11-01)
T Saito, A Wolf, N K Menon, M Saeed, C Alves, R J Bing
RÉSUMÉ
The effects of lysolecithin (lysophosphatidylcholine) derived from egg yolk as well as of synthetic lysolecithins with different aliphatic chain lengths on tension development of rabbit aortic strips were investigated. Lysolecithins caused slowly progressing, dose-dependent relaxation that was inhibited by hemoglobin, methylene blue, and nordihydroguiaretic acid. Indomethacin caused no inhibition of relaxation. The degree of relaxation was endothelium-dependent and appeared to be related to the activation of guanylate cyclase [GTP pyrophosphate-lyase (cyclizing), EC 4.6.1.2]. Superoxide dismutase failed to influence relaxation. Lysolecithins with the longest aliphatic chain were the most potent relaxants of aortic strips. The experiments suggest a role of lysolecithins through their weak detergent action on membrane dynamics of endothelial cells, resulting in the production of cyclic GMP and the relaxation of arterial smooth muscle. Lysolecithins differ in several respects from endothelium-derived relaxing factor. Endothelium-derived relaxing factor is an unstable humoral substance released from endothelium and is identical to nitric oxide, itself a labile substance causing vascular relaxation and cyclic GMP accumulation. Lysolecithins may represent a different type of endothelium-dependent muscle relaxant.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
L-α-lyso-Lecithin, Egg Yolk, Prepared enzymatically from egg yolk lecithin by treatment with phospholipase A.