Merck
  • Comparison of detergent and protease enzyme combinations for the detection of scrapie-associated fibrils from the central nervous system of sheep naturally affected with scrapie.
Comparison of detergent and protease enzyme combinations for the detection of scrapie-associated fibrils from the central nervous system of sheep naturally affected with scrapie.
Journal of comparative pathology (1997-02-01)
M J Stack, A M Aldrich, L A Davis
RÉSUMÉ
Standardized samples of tissue from the central nervous system of four sheep naturally affected with scrapie and from four healthy control sheep were subjected to a centrifugal extraction technique used to obtain scrapie-associated fibrils; the latter were then demonstrated by negative-contrast transmission electron microscopy. This regime was used to evaluate the fibril yield obtained from the 25 possible combinations of five different detergents and five different proteolytic enzymes. N-lauroylsarcosine detergent was found to be the most efficient detergent for all five enzymes, followed by sulphabetaine 3-14. Sodium dodecyl sulphate detergent was successful only in combination with a subtilisin Carlsberg enzyme. Octylglucoside and nonidet P40 detergents did not produce fibrils with any of the enzymes. Proteinase K was the least efficient of the five enzymes when used in combination with N-lauroylsarcosine; subtilisin Carlsberg, clostripain, pronase and trypsin enzymes all gave higher fibril yields. A combination of N-lauroylsarcosine detergent and subtilisin Carlsberg proteolytic enzyme gave the highest fibril yield.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
N-Lauroylsarcosine sodium salt, ≥94%