Merck
CD5L is a pleiotropic player in liver fibrosis controlling damage, fibrosis and immune cell content.
EBioMedicine (2019-05-12)
Cristina Bárcena, Gemma Aran, Luís Perea, Lucía Sanjurjo, Érica Téllez, Anna Oncins, Helena Masnou, Isabel Serra, Mónica García-Gallo, Leonor Kremer, Margarita Sala, Carolina Armengol, Pau Sancho-Bru, Maria-Rosa Sarrias
RÉSUMÉ
Chronic hepatic inflammation leads to liver fibrosis, which may progress to cirrhosis, a condition with high morbidity. Our aim was to assess the as yet unknown role of innate immunity protein CD5L in liver fibrosis. CD5L was measured by ELISA in plasma samples from cirrhotic (n = 63) and hepatitis (n = 39) patients, and healthy controls (n = 7), by immunohistochemistry in cirrhotic tissue (n = 12), and by quantitative RT-PCR in mouse liver cell subsets isolated by cell sorting. Recombinant CD5L (rCD5L) was administered into a murine model of CCl4-induced fibrosis, and damage, fibrosis and hepatic immune cell infiltration, including the LyC6hi (pro-fibrotic)-LyC6low (pro-resolutive) monocyte ratio were determined. Moreover, rCD5L was added into primary human hepatic stellate cells to study transforming growth factor β (TGFβ) activation responses. Cirrhotic patients showed elevated plasma CD5L concentrations as compared to patients with hepatitis and healthy controls (Mann-Whitney test p < 0·0001). Moreover, plasma CD5L correlated with disease progression, FIB4 fibrosis score (r:0·25, p < 0·0001) and tissue expression (r = 0·649; p = 0·022). Accordingly, CCl4-induced damage increased CD5L levels in total liver, particularly in hepatocytes and macrophages. rCD5L administration attenuated CCl4-induced injury and fibrosis as determined by reduced serum transaminase and collagen content. Moreover, rCD5L inhibited immune cell infiltration and promoted a phenotypic shift in monocytes from LyC6hi to LyC6low. Interestingly, rCD5L also had a direct effect on primary human hepatic stellate cells promoting SMAD7 expression, thus repressing TGFβ signalling. Our study identifies CD5L as a key pleiotropic inhibitor of chronic liver injury. FUND: Fundació Marató TV3, AGAUR and the ISCIII-EDRF.