Merck
Derivation of an aged mouse induced pluripotent stem cell line, IISHDOi005-A.
Stem cell research (2019-03-22)
María Del Carmen Ortuño-Costela, Victoria Cerrada, Marta García-López, Joaquín Arenas, Jesús Martínez, Alejandro Lucia, Rafael Garesse, M Esther Gallardo
RÉSUMÉ
A mouse iPSC line, IISHDOi005-A, generated from fibroblasts obtained from a mouse C57BL/6J with an age of 1 year and a half, has been obtained. For this purpose, reprogramming factors Oct3/4, Sox2, Klf4, and c-Myc were delivered using Sendai virus.